missing translation for 'onlineSavingsMsg'
Learn More

Buffer HF-Improved, Thermo Scientific Chemicals

Product Code. 11425440
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425440 1 L 1 L
11415440 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425440 Supplier Thermo Scientific Alfa Aesar Supplier No. 044627.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used For Pretreatment of Planar Silicon Devices When Plating With Nickel Plating Solution Products 44069 and 44070

Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer HF-Improved
Boiling Point 103°C to 105°C
Color Colorless
Density 1.12 g/cm3 at 20°C
Physical Form Liquid
Quantity 1 L
MDL Number MFCD00132734

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.