Learn More
Invitrogen™ Buffer Core with Cathode Bar for XCell SureLock™ Mini-Cell
Description
The Invitrogen XCell SureLock Buffer Core is a key component of the XCell SureLock Mini-Cell vertical protein gel electrophoresis system. It provides a reliable electrical interface between the upper and lower buffer chambers, ensuring consistent and reproducible gel runs.
The updated Buffer Core design incorporates a stainless-steel cathode bar, platinum wire anode electrode, gold terminals, and durable silicone gaskets, offering a precise and leak-free seal during operation. This version features enhanced robustness and improved electrical efficiency for faster gel separations.
Specifications
Specifications
| For Use With (Equipment) | SureLock Mini |
| Product Line | XCell |
| Type | Electrophoresis Core |
| For Use With | Protein Gel Electrophoresis, 1D Gel Electrophoresis, SDS PAGE, Native PAGE, IEF |
| Gel Type | Mini |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.