missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Concentrated Solution pH7.00 (Phosphate), NIST Standard for pH measurement, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
Unit Size:
1 ampoule
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 Ampule |
| MDL Number | 217672 |
| Packaging | Ampoule |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction