missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate pH 9.00, Buffer concentrate pH 9.00, Honeywell Fluka™

Product Code. 15695330 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x 1 Ea.
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15695330

Brand: Honeywell-Fluka 387486X1EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 500 ml buffer solution, sodium tetraborate / hydrochloric acid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate pH 9.00
Name Note Buffer concentrate pH 9.00
CAS Min % ≥92.0000%, ≥0.2000%
CAS 7647-01-0, 1330-43-4
CAS Max % <5.0000%
Quantity 6 x 1 Ea.
Solution Type Buffer
Packaging Ampoule
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.