missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer concentrate pH 7.00, For 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate, Honeywell Fluka™
for 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate
Brand: Honeywell Fluka 38746-6X1EA
140.27 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.02000kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer concentrate pH 7.00 | |
≥1.0000% | |
7558-79-4,7778-77-0 | |
6 x 1 Ea. | |
Ampoule |
For 500 ml buffer solution, potassium dihydrogen phosphate / disodium hydrogen phosphate | |
≤10.0000% | |
7 | |
Buffer | |
NONH for all modes of transport |