missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate, pH 3.00, citric acid / sodium hydroxide / sodium chloride solution, Honeywell Fluka™

Product Code. 15665320 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15665320 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15665320 Supplier Honeywell-Fluka Supplier No. 387421EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate
pH 3
Quantity 1 Pc.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.