missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate pH 10.00, For 500 ml buffer solution, sodium tetraborate / sodium hydroxide solution, Honeywell Fluka™

Product Code. 15605340 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15605340

Brand: Honeywell-Fluka 387491EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 500 ml buffer solution, sodium tetraborate / sodium hydroxide solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate
pH 10
Quantity 1 Pc.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.