missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer concentrate pH 10.00, For 500 ml buffer solution, sodium tetraborate / sodium hydroxide solution, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x 1 Ea.
Unit Size:
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer concentrate pH 10.00 |
| Name Note | For 500 ml buffer solution, sodium tetraborate / sodium hydroxide solution |
| Boiling Point | 100°C |
| CAS Min % | ≥90.0000%, ≥1.0000% |
| CAS Max % | ≤100.0000%, <5.0000% |
| CAS | 1330-43-4, 1310-73-2 |
| Quantity | 6 x 1 Ea. |
| Solution Type | Buffer |
| Packaging | Ampoule |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction