Learn More
Qiagen Buffer AVL (155 ml)For 250 preps: 155 ml Viral Lysis Buffer, 1550 µg Carrier RNA
Brand: Qiagen 19073
Buffer AVL (155 ml)For 250 preps: 155 ml Viral Lysis Buffer, 1550 µg Carrier RNA
Safety and Handling
- Buffer AVL (155 ml)For 250 preps: 155 ml Viral Lysis Buffer, 1550 µg Carrier RNA
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contain: guanidinium thiocyanate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.