missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, 5.00±0.02, Honeywell Fluka™ Hydrion™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
Description
- Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
Specifications
Specifications
| Chemical Name or Material | Buffer, Hydrion™ |
| pH | 5.00 ±0.02 |
| Quantity | 1 Pk. |
| MDL Number | MFCD00132752 |
| Packaging | Foil envelope |
| UN Number | NONH for all modes of transport |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction