missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Budesonide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Budesonide
Specifications
Specifications
| Chemical Name or Material | Budesonide |
| CAS | 51333-22-3 |
| Analyte or Component Names | Budesonide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H34 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Budesonide, 16α,17-[(1RS)-butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione |
| InChI Formula | InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1 |
| SMILES | CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| IUPAC Name | 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction