missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUD31 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92066-0.02ml
This item is not returnable.
View return policy
Description
BUD31 Polyclonal antibody specifically detects BUD31 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BUD31 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BUD31 homolog (S. cerevisiae), BUD31 homolog (yeast), EDG-2, EDG2MGC111202, fSAP17, functional spliceosome-associated protein 17, G10, G10 maternal transcript homolog, maternal G10 transcript, protein BUD31 homolog, Protein EDG-2, Protein G10 homolog, YCR063W | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human BUD31 (NP_003901.2). MPKVKRSRKAPPDGWELIEPTLDELDQKMREAETEPHEGKRKVESLWPIFRIHHQKTRYIFDLFYKRKAISRELYEYCIKEGYADKNLIAKWKKQGYENLCCLRCIQTRDTNFGTNCICRVPKSKLEVGRIIECTHCGCRGCSG | |
| 0.02 mL | |
| DNA replication Transcription Translation and Splicing | |
| 8896 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction