missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BUD13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558350
This item is not returnable.
View return policy
Description
Immunogen sequence: HDSPDLAPNV TYSLPRTKSG KAPERASSKT SPHWKESGAS HLSFPKNSKY EYDPDISPPR KKQAKSHFGD KKQLDSKGDC QKAT Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 61%.
BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.
Specifications
| BUD13 | |
| Polyclonal | |
| Unconjugated | |
| BUD13 | |
| AV305342; BUD13; BUD13 homolog; BUD13 homolog (yeast); Cwc26; D030060M11Rik; fSAP71; functional spliceosome-associated protein 71 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRD0 | |
| BUD13 | |
| Recombinant protein corresponding to Human BUD13. Recombinant protein control fragment (Product #RP-102827). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction