missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buclizine Hydrochloride, Mikromol™
Description
Buclizine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Buclizine Hydrochloride |
| CAS | 129-74-8 |
| Analyte or Component Names | Buclizine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H33 Cl N2 . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[[4-(1,1-dimethylethyl)phenyl]methyl]-, hydrochloride (1:2), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[[4-(1,1-dimethylethyl)phenyl]methyl]-, dihydrochloride (9CI), 1-(p-Chlorobenzhydryl)-4-(p-tert-butylbenzyl)diethylenediamine hydrochloride, 1-(p-tert-Butylbenzyl)-4-(p-chlorodiphenylmethyl)piperazine dihydrochloride, 1-p-Chlorobenzhydryl-4-p-(t)-butylbenzylpiperazine dihydrochloride, Aphilan R, Bucladin S, Buclina, Buclizine dihydrochloride, Buclizine hydrochloride, Buclodin, Histabutyzine hydrochloride, Longifene, NSC 25141, Softran, UCB 4445, Vibazine, Vibazine Hydrochloride, Piperazine, 1-(p-tert-butylbenzyl)-4-(p-chloro-alpha-phenylbenzyl)-, dihydrochloride (6CI,7CI,8CI) |
| InChI Formula | InChI=1S/C28H33ClN2.2ClH/c1-28(2,3)25-13-9-22(10-14-25)21-30-17-19-31(20-18-30)27(23-7-5-4-6-8-23)24-11-15-26(29)16-12-24;;/h4-16,27H,17-21H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CC(C)(C)c1ccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)cc1 |
| IUPAC Name | 1-[(4-tert-butylphenyl)methyl]-4-[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?