missing translation for 'onlineSavingsMsg'
Learn More

Bucladesine sodium, MedChemExpress

Product Code. 16478446
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478446 100 mg 100mg
16414477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478446 Supplier Medchem Express Supplier No. HYB0764
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.

Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.

Specifications

Chemical Name or Material Bucladesine sodium
CAS 16980-89-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H23N5NaO8P
Quantity 100 mg
Synonym Dibutyryl cAMP sodium saltDBcAMP sodium salt
Solubility Information DMSO : 170 mg/mL (345.97 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.76 mM)
SMILES O=C(CCC)O[C@H]1[C@H](N2C(N=CN=C3NC(CCC)=O)=C3N=C2)O[C@@](CO4)([H])[C@@]1([H])OP4(O[Na])=O
Molecular Weight (g/mol) 491.37
Formula Weight 491.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.