Learn More
BUCHI Reitmeyer Adapters for Rotavapor
Product Code. 10128981 Shop All Buchi Products
Change view
Click to view available options
Connection Type:
STJ 29/32
Unit Size:
Each
Product Code. Connection Type unitSize
10128981 STJ 29/32 Each
1 options
Product Code. 10128981 Supplier BUCHI Supplier No. 036576
Only null left
Add to basket
Add to basket
Bump trap adapter used to prevent foam entering the vapor duct
  • Reitmeyer style
  • Made from glass
  • SJ29/32
  • Length: 135 mm
TRUSTED_SUSTAINABILITY
Specifications
Connection Type STJ 29/32
Description Reitmeyer adapter
Material Borosilicate Glass (3.3)
Type Rotary Evaporator Adapter
For Use With (Application) Prevents contents from the evaporation flask being drawn into the condenser in the event of bumping