Learn More
BUCHI Receiving Flasks for Rotary Evaporators
Product Code. p-3649089 Shop All Buchi Products
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
3000 mL
Material (Coating):
P+G
P+G LT
Unit Size:
Each
Product Code. 10416011
Brand: BUCHI 025264
Only null left
Add to basket
Add to basket
Use these receiving flasks with any rotary evaporator. BUCHI Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.
  • Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Available in volumes from 50 mL to 3000 mL
  • Available uncoated, with P+G coating or P+G LT for dry ice condensers
Specifications
Material Borosilicate Glass (3.3)
Type Receiving Flask
For Use With (Equipment) Rotary evaporator
Includes Ball joint KS-35
Material (Coating) P+G
Shape Round
Taper 35/20
Volume (Metric) 500 mL
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis