missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Product Code. 12847695 Shop All Buchi Products
Change view
Click to view available options
Product Type:
Calibration Kit
Unit Size:
3g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12847695 Calibration Kit 3g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12847695 Supplier BUCHI Supplier No. 037454
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Vanillin, Caffeine, Phenacetin

Utilize the embedded firmware and software on your BUCHI melting point apparatus to calibrate and verify your instrument at any time. Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
TRUSTED_SUSTAINABILITY

Specifications

Product Type Calibration Kit
For Use With (Equipment) BUCHI B-540/545 Melting Point apparatus
Includes Vanillin, caffeine, phenacetin
compliance-icons compliance-icons
Product Identifier
  • Buchi B-540 melting point calibration kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: vanillin, caffeine,phenacetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.