Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.
Brand: BUCHI 11055018
Code : 61
Additional Details : CAS Number : 134-81-6 Weight : 0.10000kg
Description
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
- Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
- Guided calibrations and verifications provide easy and reliable results
- Ensures compliance from 10°C to 400°C
Specifications
Calibration Set | |
Buchi Melting Point Instrument |
Safety and Handling
- Calibration Set M-560/565
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P337+P313-If eye irritation persists: Get medical advice/attention.