missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments

Product Code. 12063303 Shop All Buchi Products
Change view
Click to view available options
Product Type:
Calibration Set
IQ/OQ Protocol
Verification Kit
For Use With (Equipment):
BUCHI Melting Point Instrument
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
12063303 Verification Kit BUCHI Melting Point Instrument Each
12053303 Calibration Set BUCHI Melting Point Instrument Each
12736400 IQ/OQ Protocol BUCHI Melting Point Instrument Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12063303 Supplier BUCHI Supplier No. 11055019
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
TRUSTED_SUSTAINABILITY

Specifications

Product Type Verification Kit
For Use With (Equipment) BUCHI Melting Point Instrument
Includes Three certified standards
compliance-icons compliance-icons
Product Identifier
  • Verification Set M-560/565
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P270-Do not eat, drink or smoke when using this product.
  • P330-Rinse mouth.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.