missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Large Capacity Evaporating Flasks

Standard glass flasks for use with various Rotavapor Evaporator models

Brand:  BUCHI 027469

Code : X3

Additional Details : Weight : 2.43000kg

 View more versions of this product

Product Code. 10523092

  • £930.00 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • Inert industrial glass (borosilicate 3.3, ISO/DIN 3585)
  • Tension relieved at 540°C

Specifications

Specifications

Standard Glass
Evaporating Flask
R-153 and R-220 rotary evaporator
10 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers