missing translation for 'onlineSavingsMsg'
Learn More

Buchi Glass column 26/920 cpl Plastic+Glas coated for preparative chromatography ID 26mm x length 920mm

Brand:  Buchi 044038

Glass column 26/920 cpl Plastic+Glas coated for preparative chromatography ID 26mm x length 920mm

Code : 62

Additional Details : Weight : 2.15000kg

Product Code. 12789475

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers