missing translation for 'onlineSavingsMsg'
Learn More

Buchi Tube glass extension 460mm x 36mm

Brand:  Buchi 017981

Tube glass extension 460mm x 36mm

Code : 62

Additional Details : Weight : 0.44000kg

Product Code. 12799525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers