missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Evaporating Flasks for Rotary Evaporators

Product Code. 10128741 Shop All Buchi Products
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Material (Coating):
Plastic and Glass
Uncoated
Taper:
29.2/32
29.2/42
Unit Size:
Each
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Material (Coating) Taper unitSize
10128741 2000 mL Plastic and Glass 29.2/32 Each
12831494 100 mL Uncoated 29.2/32 Each
10406021 250 mL Uncoated 29.2/32 Each
10280592 500 mL Uncoated 29.2/32 Each
12861494 1000 mL Uncoated 29.2/32 Each
10466201 2000 mL Uncoated 29.2/32 Each
10551172 3000 mL Uncoated 29.2/32 Each
12704963 500 mL Uncoated 29.2/42 Each
12784963 1000 mL Uncoated 29.2/42 Each
12734973 2000 mL Uncoated 29.2/42 Each
12744973 3000 mL Uncoated 29.2/42 Each
10648662 1000 mL Plastic and Glass 29.2/32 Each
10496941 500 mL Plastic and Glass 29.2/32 Each
12755113 3000 mL Plastic and Glass 29.2/32 Each
10352182 250 mL Plastic and Glass 29.2/32 Each
12778833 100 mL Plastic and Glass 29.2/32 Each
12788833 50 mL Plastic and Glass 29.2/32 Each
10178831 50 mL Uncoated 29.2/32 Each
12024592 5000 mL - 29.2/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 10128741 Supplier BUCHI Supplier No. 025323
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these evaporating flasks with any rotary evaporator. BUCHI Evaporating Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for the distillation of solvents.

  • High-performance flasks for rotary evaporators
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Constant monitoring of glass wall thickness uniformity
  • High degree of attention paid to sphericity of rotating glass parts
  • Glass parts are tension-relieved at 560°C
  • Available in volumes from 50 mL to 3000 mL
  • Available uncoated or with P+G coating
TRUSTED_SUSTAINABILITY

Specifications

Material Standard Glass
Type Evaporating Flask
For Use With (Equipment) Rotary evaporator
Material (Coating) Plastic and Glass
Shape Pear
Standard Taper Joint Number Center 29.2/32
Taper 29.2/32
Temperature (Metric) Operating 60°C
Volume (Metric) 2000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.