Learn More
BUCHI Evaporating Flask Support Ring
Product Code. 12787173 Shop All Buchi Products
Change view
Click to view available options
Material:
Silicone
Unit Size:
Each
Product Code. Material unitSize
12787173 Silicone Each
1 options
Product Code. 12787173 Supplier BUCHI Supplier No. 048618
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Flask holder for safely holding 50 to 5000mL evaporating and receiving flasks.
  • Flask ring made of non-sliding silicone
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
Type Flask Support
For Use With (Equipment) Evaporation system