missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Beaker and Drying Flasks for Rotary Evaporators

Product Code. 10446761 Shop All Buchi Products
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
1500 mL
2000 mL
Type:
Beaker Drying Flask
Beaker Flask
Powder Drying Flask
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Type unitSize
10446761 500 mL Powder Drying Flask Each
12034592 2000 mL Powder Drying Flask Each
10729782 1000 mL Powder Drying Flask Each
10091442 1500 mL Beaker Flask Each
12024782 1500 mL Beaker Drying Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10446761 Supplier BUCHI Supplier No. 000452
  • ONLINE EXCLUSIVE PRICE
    £148.00
    £134.00 / Pack of 1
    Save £14.00
    9% Off
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Special shaped flasks for large samples and powder drying

  • For use with the rotary evaporators
  • Available in multiple sizes with different standard joints
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Material Borosilicate Glass (3.3)
Type Powder Drying Flask
For Use With (Equipment) Rotary evaporator
Material (Coating) Plastic and Glass
Standard Taper Joint Side 29.2/32
Taper 29.2/32
Temperature (Metric) Operating 60°C
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.