missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BubR1 Antibody (8G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
£171.00 - £396.00
Specifications
| Antigen | BubR1 |
|---|---|
| Clone | 8G1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:2000-1:5000, Flow Cytometry 1 ug per million cells, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18039155
|
Novus Biologicals
NB100-353SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737303
|
Novus Biologicals
NB100-353 |
0.1 mL |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BubR1 Monoclonal specifically detects BubR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Specifications
| BubR1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| O60566 | |
| 701 | |
| A portion of human BubR1 tagged at the N-terminus with pMAL (residues 1-350). [UniProt# O60566] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 120 kDa |
| 8G1 | |
| Western Blot 1:2000-1:5000, Flow Cytometry 1 ug per million cells, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
| BUB1B | |
| IgG1 κ | |
| Protein G purified | |
| NB100-353 is specific for hBUBR1. There should be no cross-reactivity with hBUB1. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel