missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BubR1 Antibody (8G1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-353B
This item is not returnable.
View return policy
Description
BubR1 Monoclonal specifically detects BubR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| BubR1 | |
| Monoclonal | |
| Biotin | |
| O60566 | |
| BUB1B | |
| A portion of human BubR1 tagged at the N-terminus with pMAL (residues 1-350). [UniProt# O60566] | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-353 is specific for hBUBR1. There should be no cross-reactivity with hBUB1. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 8G1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
| Mouse | |
| 120 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators | |
| 701 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction