Learn More
Lenz Laborglasintrumente Bubble Counters
Product Code. 12653790
Click to view available options
Length (Metric):
100 mm
Unit Size:
Each
Product Code. 12653790
Brand: Lenz Laborglasintrumente 05596000
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For controlling the gas flow in distilling apparatuses
  • Height: 100 mm
  • Made of DURAN™ tubing
Specifications
Height (Metric) 100 mm
Length (Metric) 100 mm
Material Duran Glass