missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bub3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599426
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BUB3.
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| Bub3 | |
| Polyclonal | |
| Unconjugated | |
| BUB3 | |
| Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12237, 361662, 9184 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43684, Q9WVA3 | |
| BUB3 | |
| A synthesized peptide derived from human BUB3(Accession O43684), corresponding to amino acid residues S295-T328. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction