missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bub3 Polyclonal Antibody

Product Code. 12848572
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848572 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848572 Supplier Invitrogen™ Supplier No. PA520388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-20388 can be used with blocking peptide PEP-0505.

This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bub3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BUB3
Gene Accession No. O43684, Q9WVA3
Gene Alias Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5
Gene Symbols BUB3
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human bub3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12237, 9184
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.