missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bub3 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149048

Product Code. 30534687

  • £399.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bub3
Polyclonal
Unconjugated
BUB3
Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5
Goat
Antigen affinity chromatography
RUO
12237, 9184
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
O43684, Q9WVA3
BUB3
E. coli-derived recombinant human Bub3, Met1-Thr328.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.