missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bub3 Polyclonal antibody specifically detects Bub3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Bub3 |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL |
| Formulation | PBS |
| Gene Accession No. | O43684 |
| Gene Alias | BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?