missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13330-1-AP-150UL
This item is not returnable.
View return policy
Description
BUB1A is a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint.Specifications
| BUB1 | |
| Polyclonal | |
| Unconjugated | |
| BUB1 | |
| BUB1, BUB1A, BUB1L, hBUB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12235, 296137, 699 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08901, O43683 | |
| BUB1 | |
| BUB1 Fusion Protein Ag4138 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction