missing translation for 'onlineSavingsMsg'
Learn More

Bub1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18362646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362646 100 μL 100µL
18303566 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18362646 Supplier Novus Biologicals Supplier No. NBP303432100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Bub1 Polyclonal specifically detects Bub1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bub1
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BUB1 budding uninhibited by benzimidazoles 1 homolog, BUB1Amitotic checkpoint serine/threonine-protein kinase BUB1, BUB1L, budding uninhibited by benzimidazoles 1 (yeast homolog), budding uninhibited by benzimidazoles 1 homolog (yeast), EC 2.7.11.1, hBUB1mitotic spindle checkpoint kinase, putative serine/threonine-protein kinase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Bub1 (NP_004327.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 699
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.