missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bub1 Antibody (1A11C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61719-0.025ml
This item is not returnable.
View return policy
Description
Bub1 Monoclonal specifically detects Bub1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Bub1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| BUB1 budding uninhibited by benzimidazoles 1 homolog, BUB1Amitotic checkpoint serine/threonine-protein kinase BUB1, BUB1L, budding uninhibited by benzimidazoles 1 (yeast homolog), budding uninhibited by benzimidazoles 1 homolog (yeast), EC 2.7.11.1, hBUB1mitotic spindle checkpoint kinase, putative serine/threonine-protein kinase | |
| Mouse | |
| 122 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 1A11C7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BUB1 | |
| Purified recombinant fragment of human BUB1 (AA: 1-130) expressed in E. Coli. | |
| Protein G purified | |
| Breast Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Protein Kinase, Stem Cell Markers | |
| 699 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction