missing translation for 'onlineSavingsMsg'
Learn More

BU 224 hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18034092
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18034092

Brand: Tocris Bioscience 0725/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, selective I2 ligand. Putative antagonist

High affinity ligand for the imidazoline I2 binding site (Ki = 2.1 nM). Putative I2 antagonist; antagonizes the effects of imidazoline ligands on morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205437-64-5
Molecular Formula C12H12ClN3
Molecular Weight (g/mol) 233.699
InChI Key DDFHQXAQWZWRSQ-UHFFFAOYSA-N
PubChem CID 11957470
IUPAC Name 2-(4,5-dihydro-1H-imidazol-2-yl)quinoline;hydrochloride
SMILES C1CN=C(N1)C2=NC3=CC=CC=C3C=C2.Cl

Specifications

Quantity 50 mg
Synonym bu224 hydrochloride, bu 224 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride, bu224 hydrochloride, solid, 2-4,5-dihydroimidazol-2-yl-quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride 1:1
Formula Weight 233.7
Percent Purity >98%
Chemical Name or Material BU 224 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.