missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Electroporation Buffer Kit

Product Code. 15407350
Change view
Click to view available options
Quantity:
2 Bags (10 Cuvettes/Bag)
2 Bags (50 Cuvettes/Bag)
4 Bags (10 Cuvettes/Bag), 10 mL Buffer
5 Bags (10 Cuvettes/Bag)
No. of Reactions:
20
40
50
100
Unit Size:
1 set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity No. of Reactions Unit Size
15407350 2 Bags (10 Cuvettes/Bag) 20 1 set
15497340 5 Bags (10 Cuvettes/Bag) 50 1 set
15427350 2 Bags (50 Cuvettes/Bag) 100 1 set
15437350 4 Bags (10 Cuvettes/Bag), 10 mL Buffer 40 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15407350 Supplier BTX™ Supplier No. 450804
  • ONLINE EXCLUSIVE PRICE
    £656.17
    £594.81 / 1 sets
    Save £61.36
    9% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BTXPress™ Electroporation Buffer Kit offers a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

Specifications

Content And Storage 2°C to 8°C
Description BTXpress Solution Kit, 20 Reactions in 4 mm Gap Cuvettes
Cell Type Mammalian
Format Kit
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX™ and alternative electroporation systems
For Use With (Application) Transfection via Electroporation
Includes 5 mL Buffer, 20 x 4 mm Gap Cuvettes and Transfer Pipette
High-throughput Compatibility Yes
Packaging Type Kit, cuvettes individually wrapped
No. of Reactions 20
Product Line BTX Electroporation
Product Type Electroporation Buffer
Quantity 2 Bags (10 Cuvettes/Bag)
Reagent Type Universal Electroporation Buffer
Sterility Sterile
Volume (Metric) 5 mL
Show More Show Less
compliance-icons
Product Identifier
  • BTXpress electroporation solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Supplemental information
  • MIXTURE LIST-Contain: trade secret
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.