missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Electroporation Buffer

Product Code. p-7222370
Click to view available options
Quantity:
2 mL
5 mL
10 mL
No. of Reactions:
20
50
100
Unit Size:
10mL
2mL
5mL
This item is not returnable. View return policy

Product Code. 15477340

Brand: BTX™ 450801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BTXPress™ Electroporation Buffer is a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

Specifications

Buffer Universal Electroporation Buffer
Content And Storage 2°C to 8°C
Color Clear
Description BTXpress Solution, 2.0 ml Bottle for up to 20 Reactions
Cell Type Mammalian
Format Bottle
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX™ and other electroporators
For Use With (Application) Transfection via Electroporation
Includes 2 mL Buffer only
Packaging Type Bottle
No. of Reactions 20
Product Line BTX Electroporation
Product Type Electroporation Buffer
Quantity 2 mL
Sterility Sterile
Volume (Metric) 2 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.