missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Two-Needle Array™ Electrodes: Handle Only

Product Code. 15477290
Click to view available options
Size (Metric) Gap:
5 mm
10 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15477290

Brand: BTX™ 450206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

8cm long array handle
s

Handle for use with BTX Two-Needle Array™ Electrodes (cat. no. BTX531).

  • Reusable
  • 45-0167 handle designed for 45-0120 10 mm needle tips
    • Used for larger muscle masses
  • 45-0168 handle designed for 45-0121 5 mm needle tips
    • Recommended for smaller muscle masses

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
For Use With (Equipment) 2-needle Array (45-0168), ECM 830, 2001, 2001+ and Gemini X2 Generators
Material Delrin
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 500 V
Description 2-Needle Array Handle, 5 mm
Includes Handle only. Requires 45-0168
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Size (English) Gap 0.19 in.
Size (Metric) Gap 5 mm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Required Accessories 2-needle Array (45-0168)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.