missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes
Two-Needle Array™ Electrodes designed for in vivo drug or gene delivery.
Brand: BTX™ 450121
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- Sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
Medical Grade Stainless Steel | |
Electroporation Accessory | |
2-Needle Array, 5 mm Gap, Pkg. of 6 | |
Pk. 6 Needle Array. Handle and connectors required | |
20 mm | |
6 Pk. | |
5 mm | |
Banana Plug to Banana Plug | |
Handle, Connectors |
ECM 830, ECM 2001+, ECM 630, Legacy ECM 2001 Generator | |
BTX Electroporation | |
500 V | |
Straight/Needle | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
0.19 in. | |
Sterile | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.