missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrode Kit
Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.
£530.00 - £580.00
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Muscle Gene Therapy |
---|---|
Material | Stainless Steel, Delrin |
Product Line | BTX Electroporation |
Voltage | 500 V |
Length (English) Electrode | 0.78 in. |
Includes:
- Handle
- Six two-needle arrays
Product Code | Brand | Size (Metric) Gap | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Gap | Price | Quantity & Availability | |||||
15407290
|
BTX™
450167 |
10 mm |
£580.00
Each |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15417290
|
BTX™
450168 |
5 mm |
£530.00
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Consists of reusable array handle and sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
- Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
BTX Electroporation | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
Non-sterile | |
Square Wave |
Stainless Steel, Delrin | |
500 V | |
20 mm | |
5°C to 40°C | |
Banana Cables |
Intended for Research use only. Not for use in diagnostic or clinical procedures.