missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genetrodes™ Electrodes
Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.
£484.00 - £540.00
Specifications
For Use With (Application) | In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery |
---|---|
Material | Gold |
Voltage | 200 V |
Includes | Genetrode only |
Life Span | ∼1500+ Pulses |
Product Code | Brand | Electrode Shape | Length (English) Electrode | Length (Metric) Electrode | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Electrode Shape | Length (English) Electrode | Length (Metric) Electrode | Price | Quantity & Availability | |||
15457260
|
BTX™
450116 |
Bent L-shaped | 0.11 in. | 3 mm |
£484.00
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15447260
|
BTX™
450115 |
Bent L-shaped | 0.19 in. | 5 mm |
£540.00
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15427260
|
BTX™
450113 |
Straight | 0.19 in. | 5 mm |
£484.00
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15467260
|
BTX™
450117 |
Bent L-shaped | 0.039 in. | 1 mm |
£484.00
Each |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15437260
|
BTX™
450114 |
Straight | 0.39 in. | 10 mm |
£484.00
Each |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Specifications
In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
200 V | |
∼1500+ Pulses | |
10 μsec. to 99 msec. in PBS | |
0.03 to 0.39 in. | |
Gold Tip | |
Banana Plug to Micrograbber |
Gold | |
Genetrode only | |
1 sec. to 99 msec. | |
1 Ea. | |
1 to 10 mm | |
Non-sterile | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.