missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Caliper Electrodes

Caliper Electrodes for in-vivo electroporation.

Brand:  BTX™ 450102

 View more versions of this product

Product Code. 15437250

  • £1165.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For in vivo electroporation, specifically the delivery of molecules or drugs into a desired tissue
  • Consist of two stainless-steel electrodes that grasp the surface site to be electroporated
  • Applications include:
    • Electrochemotherapy (ECT): the use of pulsed electric fields to deliver chemotherapeutic drugs into cancerous tumors or
    • Transdermal Delivery (TDD): use of a pulsed electric field to deliver drugs or other molecules through the skin to the targeted site
  • For use with BTX Gemini X2 electroporation systems.
Specifications

Specifications

In-Vivo Drug or Gene Delivery, Electrochemotherapy, Transdermal Applications, Intact Plant Applications
Stainless Steel
Electroporation accessory
Caliper Electrode, 2.0 cm x 2.0 cm with extra 1.5 cm x 1.5 cm electrodes included, Stainless Steel
Caliper Electrode, 2.0 cm x 2.0 cm with extra 1.5 cm x 1.5 cm electrode included
2 cm
10 μsec. to 99 msec.
0.039 to 5.11 in.
Stainless Steel Caliper
2 cm
Banana Cables
ECM 830, Gemini X2, ECM 2001+, and Legacy ECM 2001
BTX Electroporators
500 V
Square
0.79 in.
Unlimited when properly maintained
2 Pk.
1 to 130 mm
0.79 in.
Non-sterile
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Intended for Research use only. Not for use in diagnostic or clinical procedures.