missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Caliper Electrodes
Caliper Electrodes for in-vivo electroporation.
Brand: BTX™ 450101
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- For in vivo electroporation, specifically the delivery of molecules or drugs into a desired tissue
- Consist of two stainless-steel electrodes that grasp the surface site to be electroporated
- Applications include:
- Electrochemotherapy (ECT): the use of pulsed electric fields to deliver chemotherapeutic drugs into cancerous tumors or
- Transdermal Delivery (TDD): use of a pulsed electric field to deliver drugs or other molecules through the skin to the targeted site
- For use with BTX Gemini X2 electroporation systems.
Specifications
In-Vivo Drug or Gene Delivery, Transdermal Applications, Intact Plant Applications, Electrochemotherapy | |
Brass | |
Caliper Electrode , 1.0 cm X 1.0 cm, Brass | |
0.39 in. | |
Unlimited when properly maintained | |
1 Ea. | |
1 to 130 mm | |
0.39 in. | |
Non-sterile | |
Exponential Decay Wave, Square Wave |
ECM630, ECM 830, Gemini X2, ECM 2001+, and Legacy ECM 2001 Electroporation Systems | |
500 V | |
Square | |
1 cm | |
10 μsec. to 99 msec. | |
0.039 to 5.11 in. | |
Brass, square | |
1 cm | |
Banana Cables |
Intended for Research use only. Not for use in diagnostic or clinical procedures.