missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode

Product Code. 15487320
Change view
Click to view available options
Diameter (Metric):
1 mm
3 mm
5 mm
7 mm
10 mm
Material:
Platinum
Stainless Steel
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Material unitSize
15487320 1 mm Platinum Each
15497320 3 mm Platinum Each
15417330 5 mm Platinum Each
15407330 7 mm Platinum Each
15477260 7 mm Stainless Steel Each
15487260 10 mm Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15487320 Supplier BTX™ Supplier No. 450486
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

For Use With (Application) Vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
For Use With (Equipment) BTX 830, ECM 2001+, Gemini X2 Generators
Material Platinum
Product Line BTX Electroporation
Product Type Tweezer Style Electrode
Voltage 200 V
Description Platinum Tweezertrode, 1 mm Diameter
Diameter (English) 0.039 in.
Diameter (Metric) 1 mm
Electrode Shape Circular
Length (English) Electrode 0.47 in.
Length (Metric) Electrode 11.9 mm
Operating Temperature (English) 41°F to 104°F
Pulse Number Range 1 sec. to 99 msec.
Pulse Length Range 1 μsec. to 200 msec.
Operating Temperature (Metric) 5°C to 40°C
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (Metric) Gap Adjustable: < 1 mm to > 2 cm
Tip Type Tweezer Style
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.