missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genepaddles™ Electroporation Electrodes
Genepaddles™ Electroporation Electrodes are paddle-style reusable electrodes suitable for both in vitro and in vivo gene delivery.
£555.00 - £1340.00
Specifications
For Use With (Application) | In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery |
---|---|
For Use With (Equipment) | ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 |
Material | Gold-plated |
Voltage | 200 V |
Pulse Length Range | 10 μsec. to 99 msec. in PBS |
Product Code | Brand | Product Type | Length (Metric) | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Length (Metric) | Length (English) | Price | Quantity & Availability | |||
15437290
|
BTX™
450170 |
Genepaddles Kit | 5 mm | 0.19 in. |
£1340.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15427270
|
BTX™
450123 |
Genepaddles | 5 mm | 0.19 in. |
£595.00
Each |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15417270
|
BTX™
450122 |
Genepaddles | 3 mm | 0.11 in. |
£555.00
Each |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15427290
|
BTX™
450169 |
Genepaddles Kit | 3 mm | 0.11 in. |
£1315.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
- Two models to suit the size of the target area
- Gold plated, noninvasive
- Thickness: 1 mm
- Easily cleaned
- Life span is approximately 1500 pulses
- 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)
Specifications
In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery | |
Gold-plated | |
10 μsec. to 99 msec. in PBS | |
0.03 to 0.39 in. | |
Non-sterile | |
Square Wave |
ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 | |
200 V | |
1 Ea. | |
1 to 10 mm | |
Banana Plug to Micrograbber |
Intended for Research use only. Not for use in diagnostic or clinical procedures.