missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: EDWESKQMPQ YRGRTEFVKD SIAGGRVSLR LKNITPSDIG LYGCWFSSQI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 77%; Horse: 77%; Human: 100%; Pig: 77%.
BTNL3 is expressed in small intestine, colon, testis, spleen, and leukocyte and plays a role in lipid metabolism. Two isoforms have been described.
Specifications
Specifications
| Antigen | BTNL3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | BTNL3 |
| Gene Accession No. | Q6UXE8 |
| Gene Alias | BTN9.1; BTNL3; BTNLR; butyrophilin like 3; butyrophilin-like 3; butyrophilin-like protein 3; Butyrophilin-like receptor; COLF4100; UNQ744/PRO1472 |
| Gene Symbols | BTNL3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
