missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTNL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25110-1-AP-150UL
This item is not returnable.
View return policy
Description
BTNL2 (butyrophilin-like protein 2), also known as BTL-II, is a 455 amino acid single-pass type II membrane protein that acts as a negative regulator of T cell proliferation. Expressed in thymus, brain, kidney, heart, liver, small intestine, testis, ovary, pancreas and leukocytes, BTNL2 exists as at least six alternatively spliced isoforms, which are encoded by a gene located on human chromosome 6. Expression of BTNL2 isoform 3 is associated with susceptibility to sarcoidosis type 2 (SS2), an immune disorder that causes chronic inflammatory granulomatous lesions.Specifications
| BTNL2 | |
| Polyclonal | |
| Unconjugated | |
| BTNL2 | |
| BTL II, BTNL2, Butyrophilin like protein 2, HSBLMHC1, SS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 547431, 56244 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70355, Q9UIR0 | |
| Btnl2 | |
| BTNL2 Fusion Protein Ag18803 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction