missing translation for 'onlineSavingsMsg'
Learn More

BTN3A3 Antibody, Novus Biologicals™

Product Code. 18324779 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324779 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324779 Supplier Novus Biologicals Supplier No. H00010384B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

BTN3A3 Polyclonal antibody specifically detects BTN3A3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN3A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH15815
Gene Alias BTF3butyrophilin 3, butyrophilin subfamily 3 member A3, butyrophilin, subfamily 3, member A3
Host Species Mouse
Immunogen BTN3A3 (AAH15815, 1 a.a. - 584 a.a.) fullAAH15815length human protein. MKMASSLAFLLLNFHVSLFLVQLLTPCSAQFSVLGPSGPILAMVGEDADLPCHLFPTMSAETMELRWVSSSLRQVVNVYADGKEVEDRQSAPYRGRTSILRDGITAGKAALRIHNVTASDSGKYLCYFQDGDFYEKALVELKVAALGSDLHIEVKGYEDGGIHLECRSTGWYPQPQIKWSDTKGENIPAVEAPVVADGVGLYAVAASVIMRGSSGGGVSCIIRNSLLGLEKTASISIADPFFRSAQPWIAALAGTLPISLLLLAGASYFLWRQQKEKIALSRETEREREMKEMGYAATEQEISLREKLQEELKWRKIQYMARGEKSLAYHEWKMALFKPADVILDPDTANAILLVSEDQRSVQRAEEPRDLPDNPERFEWRYCVLGCENFTSGRHYWEVEVGDRKEWHIGVCSKNVERKKGWVKMTPENGYWTMGLTDGNKYRALTEPRTNLKLPEPPRKVGIFLDYETGEISFYNATDGSHIYTFPHASFSEPLYPVFRILTLEPTALTICPIPKEVESSPDPDLVPDHSLETPLTPGLANESGEPQAEVTSLLLPAHPGAEVSPSATTNQNHKLQARTEALY
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 10384
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.